![N,N′-Diacetylchitobiose ≥96% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/365/165/f5adb04f-9d5c-4eb0-9576-d4261459fb5c/640/f5adb04f-9d5c-4eb0-9576-d4261459fb5c.png)
N,N′-Diacetylchitobiose
![N,N′-Diacetylchitobiose ≥96% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/365/165/f5adb04f-9d5c-4eb0-9576-d4261459fb5c/640/f5adb04f-9d5c-4eb0-9576-d4261459fb5c.png)
Synonym(s):
2-Acetamido-2-deoxy-4-O-(2-acetamido-2-deoxy-β-D-glucopyranosyl)-D-glucopyranose, 4-O-(2-Acetamido-2-deoxy-β-D-glucopyranosyl)-2-acetamido-2-deoxy-D-glucose, Chitobiose
Empirical Formula (Hill Notation):
C16H28N2O11
CAS Number:
Molecular Weight:
424.40
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing