
N-Desalkylflurazepam

Synonym(s):
Norfludiazepam, 7-Chloro-5-(2-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Desalkylflurazepam
Empirical Formula (Hill Notation):
C15H10ClFN2O
CAS Number:
Molecular Weight:
288.70
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing