
Monoclonal Anti-PD-1 antibody produced in mouse

Synonym(s):
Anti-CD279, Anti-PDCD-1, Anti-Programmed cell death 1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing