![Monoclonal Anti-GRN antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL5695, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/312/928/1e3c6b78-b502-4022-8502-9a36d15181e5/640/1e3c6b78-b502-4022-8502-9a36d15181e5.jpg)
Monoclonal Anti-GRN antibody produced in mouse
![Monoclonal Anti-GRN antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL5695, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/312/928/1e3c6b78-b502-4022-8502-9a36d15181e5/640/1e3c6b78-b502-4022-8502-9a36d15181e5.jpg)
Synonym(s):
CLN11, PCDGF, PGRN
Products
Product Category
Brand
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing