![Metformin Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/209/526/a077f827-9636-4aef-b148-c40baa4aad1e/640/a077f827-9636-4aef-b148-c40baa4aad1e.jpg)
Metformin Related Compound B
![Metformin Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/209/526/a077f827-9636-4aef-b148-c40baa4aad1e/640/a077f827-9636-4aef-b148-c40baa4aad1e.jpg)
Synonym(s):
1-Methylbiguanide monohydrochloride
Empirical Formula (Hill Notation):
C3H9N5 · HCl
CAS Number:
Molecular Weight:
151.60
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2