
Levalbuterol Related Compound F

Synonym(s):
(1R)-1-(4-(Benzyloxy)-3-(hydroxymethyl)phenyl)-2-(tert-butylamino)ethan-1-ol, (R)-α-{[(1,1-Dimethylethyl)amino]methyl}-4-(phenylmethoxy)-1,3-benzenedimethanol, (R)-4-Benzyl albuterol
Empirical Formula (Hill Notation):
C20H27NO3
CAS Number:
Molecular Weight:
329.43
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2