Skip to Content
Merck
L-(+)-Erythrulose ≥75% (HPLC)

L-(+)-Erythrulose

L-(+)-Erythrulose ≥75% (HPLC)
Synonym(s):
S-1,3,4-Trihydroxy-2-butanone, L-Glycero-2-tetrulose
Empirical Formula (Hill Notation):
C4H8O4
CAS Number:
Molecular Weight:
120.10

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing