![Imiquimod Related Compound D pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/916/149/a806b4ea-ecc5-4915-98f2-5dfc7e9a71dd/640/a806b4ea-ecc5-4915-98f2-5dfc7e9a71dd.jpg)
Imiquimod Related Compound D
![Imiquimod Related Compound D pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/916/149/a806b4ea-ecc5-4915-98f2-5dfc7e9a71dd/640/a806b4ea-ecc5-4915-98f2-5dfc7e9a71dd.jpg)
Synonym(s):
1-Propyl-1H-imidazo[4,5-c]quinolin-4-amine
Empirical Formula (Hill Notation):
C13H14N4
CAS Number:
Molecular Weight:
226.28
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2