![Ethyl-beta-D-glucuronide solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/160/765/c4882448-295d-4a27-a709-c3839f307eb3/640/c4882448-295d-4a27-a709-c3839f307eb3.png)
Ethyl-beta-D-glucuronide solution
![Ethyl-beta-D-glucuronide solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/160/765/c4882448-295d-4a27-a709-c3839f307eb3/640/c4882448-295d-4a27-a709-c3839f307eb3.png)
Synonym(s):
Ethyl-β-D-glucuronide
Empirical Formula (Hill Notation):
C8H14O7
CAS Number:
Molecular Weight:
222.19
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing