![DMT-2′Fluoro-dU Phosphoramidite configured for PerkinElmer, configured for Polygen](/deepweb/assets/sigmaaldrich/product/structures/154/407/9e93e0ee-0666-4d80-883f-9921ca7da402/640/9e93e0ee-0666-4d80-883f-9921ca7da402.png)
DMT-2′Fluoro-dU Phosphoramidite
![DMT-2′Fluoro-dU Phosphoramidite configured for PerkinElmer, configured for Polygen](/deepweb/assets/sigmaaldrich/product/structures/154/407/9e93e0ee-0666-4d80-883f-9921ca7da402/640/9e93e0ee-0666-4d80-883f-9921ca7da402.png)
Synonym(s):
DMT-2′-Fluoro-dU amidite
Empirical Formula (Hill Notation):
C39H46FN4O8P
CAS Number:
Molecular Weight:
748.78
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing