Skip to Content
Merck
Dibutyltin dichloride 96%

Dibutyltin dichloride

Dibutyltin dichloride 96%
Synonym(s):
Dibutyldichlorotin, Dibutyltin dichloride
Linear Formula:
(CH3CH2CH2CH2)2SnCl2
CAS Number:
Molecular Weight:
303.84

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing