Skip to Content
Merck
Dibutyltin diacetate technical grade

Dibutyltin diacetate

Dibutyltin diacetate technical grade
Synonym(s):
Diacetoxydibutyltin
Linear Formula:
(CH3CH2CH2CH2)2Sn(OCOCH3)2
CAS Number:
Molecular Weight:
351.03
EC Number:

Products

Shipping
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing