
Cefaclor, Delta-3 isomer

Synonym(s):
Delta-3-cefaclor, (2R,6R,7R)- and (2S,6R,7R)-7-{[(2R)-2-Amino-2-phenylacetyl]amino}-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid
Empirical Formula (Hill Notation):
C15H14ClN3O4S
CAS Number:
Molecular Weight:
367.81
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing