![Bis(2-ethylhexyl) amine for synthesis](/deepweb/assets/sigmaaldrich/product/images/234/177/871f9945-0840-4395-8732-4fde66e3d44f/640/871f9945-0840-4395-8732-4fde66e3d44f.jpg)
Bis(2-ethylhexyl) amine
![Bis(2-ethylhexyl) amine for synthesis](/deepweb/assets/sigmaaldrich/product/images/234/177/871f9945-0840-4395-8732-4fde66e3d44f/640/871f9945-0840-4395-8732-4fde66e3d44f.jpg)
Synonym(s):
Bis(2-ethylhexyl) amine
Linear Formula:
[CH3(CH2)3CH(C2H5)CH2]2NH
CAS Number:
Molecular Weight:
241.46
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing