Anti-GALNT1 antibody produced in rabbit
Synonym(s):
Anti-GalNAc-T1, Anti-Polypeptide GalNAc transferase 1, Anti-Polypeptide N-acetylgalactosaminyltransferase 1, Anti-Protein-UDP acetylgalactosaminyltransferase 1, Anti-UDP- GalNAc:polypeptide N-acetylgalactosaminyltransferase 1, Anti-pp-GaNTase 1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1