Anti-DNPEP antibody produced in rabbit
Synonym(s):
Anti-ASPEP, Anti-Aspartyl Aminopeptidase, Anti-DAP, Anti-Aspartyl aminopeptidase
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing