
Anti-COX11 antibody produced in rabbit

Synonym(s):
Anti-COX11 cytochrome c oxidase assembly homolog (yeast), Anti-COX11P
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1