
Anti-CLN6 antibody produced in rabbit

Synonym(s):
Ceroid-lipofuscinosis neuronal protein 6, Protein CLN6, Anti-FLJ20561, Anti-HsT18960, Anti-nclf
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing