
Anti-CEP55 antibody produced in rabbit

Synonym(s):
Anti-C10orf3, Anti-CT111, Anti-Centrosomal protein 55kDa, Anti-FLJ10540
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing