
Anti-BRIP1 antibody produced in rabbit

Synonym(s):
Anti-BACH1, Anti-BRCA1 interacting protein C-terminal helicase 1, Anti-FANCJ, Anti-OF
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1