![Acetobromo-α-D-galactose ≥93% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/314/243/613cdf1b-b68d-497b-9ee2-943c7450cf37/640/613cdf1b-b68d-497b-9ee2-943c7450cf37.png)
Acetobromo-α-D-galactose
![Acetobromo-α-D-galactose ≥93% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/314/243/613cdf1b-b68d-497b-9ee2-943c7450cf37/640/613cdf1b-b68d-497b-9ee2-943c7450cf37.png)
Synonym(s):
2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide
Empirical Formula (Hill Notation):
C14H19BrO9
CAS Number:
Molecular Weight:
411.20
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing