Skip to Content
Merck
6-Acetyl-1,1,2,4,4,7-hexamethyltetralin analytical standard

6-Acetyl-1,1,2,4,4,7-hexamethyltetralin

6-Acetyl-1,1,2,4,4,7-hexamethyltetralin analytical standard
Synonym(s):
1,2,3,4-Tetrahydro-1,1,3,4,4,6-hexamethyl-7-naphthyl methyl ketone, 1-(5,6,7,8-Tetrahydro-3,5,5,6,8,8-hexamethyl-2-naphthalenyl)ethanone, 5′,6′,7′,8′-Tetrahydro-3′,5′,5′,6′,8′,8′-hexamethyl-2′-acetonaphthone, 6-Acetyl-1,1,2,4,4,7-hexamethyl-1,2,3,4-tetrahydronaphthalene, AHMT, Fixolide, Musk tonalid, NSC 19550
Empirical Formula (Hill Notation):
C18H26O
CAS Number:
Molecular Weight:
258.40
EC Number:

Products

Shipping
Formula Weight
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing