![4-Nitrophenyl octanoate ≥90.0% (GC)](/deepweb/assets/sigmaaldrich/product/structures/421/158/bdc4c72f-6341-4fe3-8008-5ac7d21df87e/640/bdc4c72f-6341-4fe3-8008-5ac7d21df87e.png)
4-Nitrophenyl octanoate
![4-Nitrophenyl octanoate ≥90.0% (GC)](/deepweb/assets/sigmaaldrich/product/structures/421/158/bdc4c72f-6341-4fe3-8008-5ac7d21df87e/640/bdc4c72f-6341-4fe3-8008-5ac7d21df87e.png)
Synonym(s):
4-Nitrophenyl caprylate, Caprylic acid 4-nitrophenyl ester, Octanoic acid 4-nitrophenyl ester
Empirical Formula (Hill Notation):
C14H19NO4
CAS Number:
Molecular Weight:
265.30
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing