Skip to Content
Merck
2-Furoylglycine ≥98%

2-Furoylglycine

2-Furoylglycine ≥98%
Synonym(s):
N-(2-Furoyl)glycine
Empirical Formula (Hill Notation):
C7H7NO4
CAS Number:
Molecular Weight:
169.13

Products

Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing