
25-Hydroxyvitamin D2 (6,19,19-d3) solution

Synonym(s):
25-Hydroxycalciferol (6,19,19-d3) solution, 25-Hydroxyergocalciferol (6,19,19-d3) solution
Empirical Formula (Hill Notation):
C28D3H41O2
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-2 of 2