
1,3,6-Tri-O-galloyl-β-D-glucose

Synonym(s):
β-D-Glucopyranose 1,3,6-trigallate, β-D-Glucopyranose-1,3,6-tris(3,4,5-trihydroxybenzoate), 1,3,6-Tri-O-galloyl-β-D-glucopyranose, NP 005114, NSC 69861
Empirical Formula (Hill Notation):
C27H24O18
CAS Number:
Molecular Weight:
636.47
Products
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing