Skip to Content
Merck
All Photos(1)

Key Documents

SML3431

Sigma-Aldrich

JBSNF-000088

≥98% (HPLC)

Synonym(s):

6-Methoxy-3-pyridinecarboxamide, 6-Methoxyniacinamide, 6-Methoxynicotinamide, NSC 70628

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O2
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C7H8N2O2/c1-11-6-3-2-5(4-9-6)7(8)10/h2-4H,1H3,(H2,8,10)

InChI key

KXDSMFBEVSJYRF-UHFFFAOYSA-N

Biochem/physiol Actions

JBSNF-000088 is a cell penetrant, potent and selective inhibitor of Nicotinamide N-methyltransferase (NNMT) that reduces 1-methyl-nicotinamide (MNA) and improved glucose control in mice models of metabolic disease. JBSNF-000088 potently inhibits growth of PFA ependymoma cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service