Skip to Content
Merck
All Photos(1)

Key Documents

719730

Sigma-Aldrich

5-(Tributylstannyl)pyrimidine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30N2Sn
CAS Number:
Molecular Weight:
369.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.515

bp

134-138 °C/2 mmHg

density

1.117 g/mL at 25 °C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1cncnc1

InChI

1S/C4H3N2.3C4H9.Sn/c1-2-5-4-6-3-1;3*1-3-4-2;/h2-4H;3*1,3-4H2,2H3;

InChI key

QRDQHTJNKPXXRQ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service