Merck
  • Inhibitory effect of silver nanoparticles on proliferation of estrogen-dependent MCF-7/BUS human breast cancer cells induced by butyl paraben or di-n-butyl phthalate.
Inhibitory effect of silver nanoparticles on proliferation of estrogen-dependent MCF-7/BUS human breast cancer cells induced by butyl paraben or di-n-butyl phthalate.
Toxicology and applied pharmacology (2017-10-28)
Joanna Roszak, Anna Smok-Pieniążek, Katarzyna Domeradzka-Gajda, Jarosław Grobelny, Emilia Tomaszewska, Katarzyna Ranoszek-Soliwoda, Grzegorz Celichowski, Maciej Stępnik
ABSTRACT
In this study the effect of silver nanoparticles (AgNPs) on proliferation of estrogen receptor (ER)-positive human breast cancer MCF-7/BUS cells was assessed by means of in vitro assay. The cells were exposed in the absence of estrogens to AgNPs alone or in combination with aluminum chloride (AlCl
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture