Merck
  • Sinonasal Delivery of Resveratrol via Mucoadhesive Nanostructured Microparticles in a Nasal Polyp Mouse Model.
Sinonasal Delivery of Resveratrol via Mucoadhesive Nanostructured Microparticles in a Nasal Polyp Mouse Model.
Scientific reports (2017-01-11)
Mingyu Lee, Chun Gwon Park, Beom Kang Huh, Se-Na Kim, Seung Ho Lee, Roza Khalmuratova, Jong-Wan Park, Hyun-Woo Shin, Young Bin Choy
ABSTRACT
Resveratrol (RSV) has been shown to effectively suppress chronic rhinosinusitis with nasal polyps in a mouse model; however, when locally administered to the sinonasal cavity, bolus RSV is limited by low drug bioavailability owing to its low aqueous solubility and relatively rapid clearance from the administration site. To address this limitation, we propose mucoadhesive nanostructured microparticles (PLGA/PEG NM) as a potential carrier for the sinonasal delivery of RSV. In this study, PLGA/PEG NM released RSV in a sustained manner. Owing to the enlarged specific surface area of the nanostructures, PLGA/PEG NM had synergistically enhanced mucoadhesiveness and thus showed improved in vivo retention properties in the sinonasal cavity. Therefore, when tested in a mouse nasal polyp model, PLGA/PEG NM mitigated polyp formation and restored epithelial integrity better than the control treatments. The therapeutic effect was similar at half the dose of PLGA/PEG NM, suggesting improved local bioavailability of RSV in the sinonasal cavity.
Sigma-Aldrich
3,3′-Diethylthiatricarbocyanine iodide, 99%
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)