Merck
  • Use of solid-phase microextraction in the investigation of chemical communication in social wasps.
Use of solid-phase microextraction in the investigation of chemical communication in social wasps.
Journal of chromatography. A (2000-04-11)
M F Sledge, G Moneti, G Pieraccini, S Turillazzi
ABSTRACT
Solid-phase microextraction has been used to investigate chemical communication in several social wasp species. Using the technique to analyse exocrine gland secretions, we demonstrate that the results are comparable with those obtained with the more classical methods that use solvents, eliminating, in many cases, the shortcomings of these methods in insect pheromone analysis. As a result of its simplicity this technique is very suitable for research on the chemical ecology of social wasps, and on insect communication in general.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder