Merck
  • Determination of nitrazepam and its main metabolites in urine by thin-layer chromatography and direct densitometry.
Determination of nitrazepam and its main metabolites in urine by thin-layer chromatography and direct densitometry.
Journal of chromatography (1985-04-12)
T Inoue, T Niwaguchi
ABSTRACT
A method for the direct quantitative densitometry of nitrazepam and its main metabolites (7-aminonitrazepam, 7-acetamidonitrazepam and 2-amino-5-nitrobenzophenone) in urine was developed. The unchanged drug and its metabolites were extracted with benzene-dichloromethane (4:1), subjected to thin-layer chromatography, and determined by direct ultraviolet densitometry. Recovery experiments showed that the method was quantitative. The limit of detection was 5 ng/ml for 2-amino-5-nitrobenzophenone and 10 ng/ml for other compounds. The method was applied to the determination of nitrazepam and its metabolites excreted in human urine after administration of 10 mg of the drug.
Product Description
Supelco
7-Aminonitrazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2-Amino-5-nitrobenzophenone, ≥98%