Merck
  • Embossing of organic thin films using a surfactant assisted lift-off technique.
Embossing of organic thin films using a surfactant assisted lift-off technique.
Journal of colloid and interface science (2012-09-15)
Xin Zhang, Gengxin Zhang, Yen-Chih Liao, Brandon L Weeks, Zhao Zhang
ABSTRACT
A simple technique for patterning organic materials using a surfactant assisted lift-off method is proposed. Thin films of various organic materials are prepared, and areas in contact with a surfactant coated poly(dimethylsiloxane) (PDMS) stamp are selectively removed. The general applicability of this technique is shown for materials containing nitrate, amine, and carboxylic acid functional groups. This technique provides a new methodology for fabricating patterns with vertical dimensions ranging from 30 nm up to 3 μm on organic thin films with specific functional groups.
Product Description
Supelco
Pentaerythritol tetranitrate solution, 1000 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Pentaerythritol tetranitrate solution, 10 mg/mL in acetonitrile, ampule of 5 mL, certified reference material, Cerilliant®