Merck
  • Newly detected specific hydrogenation of the conjugated double bond of unsaturated alkaloid lactones by Aspergillus sp.
Newly detected specific hydrogenation of the conjugated double bond of unsaturated alkaloid lactones by Aspergillus sp.
Biotechnology letters (2005-09-15)
Hong Guan, Song You, Li Yang, Xu Wang, Rui Ni
ABSTRACT
A new isolate of Aspergillus sp. hydrogenated the gamma,delta-double bond of securinine (143 mg l(-1)) to give 14,15-dihydrosecurinine at over 98% (w/w) yield after 8 h. It also hydrogenated the C11(13) double bond of 3-hydroxy-1(10),3,11(13)-guaiatriene-12,6-olide-2-one (HGT) (200 mg l(-1)) to give 3-hydroxy-1(10),3-guaiadiene-12,6-olide-2-one with over 98% (w/w) conversion after 24 h.
Sigma-Aldrich
Securinine, ≥98% (HPLC)