Merck
  • Silver acetate catalyzed hydroamination of 1-(2-(sulfonylamino)phenyl)prop-2-yn-1-ols to (Z)-2-methylene-1-sulfonylindolin-3-ols.
Silver acetate catalyzed hydroamination of 1-(2-(sulfonylamino)phenyl)prop-2-yn-1-ols to (Z)-2-methylene-1-sulfonylindolin-3-ols.
The Journal of organic chemistry (2012-08-17)
Dewi Susanti, Fujiet Koh, Jeffrey Antonius Kusuma, Prasath Kothandaraman, Philip Wai Hong Chan
ABSTRACT
A method to prepare (Z)-2-methylene-1-sulfonylindolin-3-ols efficiently that relies on silver acetate catalyzed hydroamination of 1-(2-(sulfonylamino)phenyl)prop-2-yn-1-ols is reported. The reactions proceed rapidly at room temperature with catalyst loadings as low as 1 mol % under conditions that did not require the exclusion of air or moisture. The utility of this N-heterocyclic ring-forming strategy as a synthetic tool that makes use of unsaturated alcohols was exemplified by the conversion of the (Z)-2-methylene-1-sulfonylindolin-3-ol to examples of other members of the indole family of compounds.
Sigma-Aldrich
Silver acetate, 99.99% trace metals basis
Sigma-Aldrich
Silver acetate, ReagentPlus®, 99%