Merck
  • Antimetastatic potential of amide-linked local anesthetics: inhibition of lung adenocarcinoma cell migration and inflammatory Src signaling independent of sodium channel blockade.
Antimetastatic potential of amide-linked local anesthetics: inhibition of lung adenocarcinoma cell migration and inflammatory Src signaling independent of sodium channel blockade.
Anesthesiology (2012-08-01)
Tobias Piegeler, E Gina Votta-Velis, Guoquan Liu, Aaron T Place, David E Schwartz, Beatrice Beck-Schimmer, Richard D Minshall, Alain Borgeat
ABSTRACT
Retrospective analysis of patients undergoing cancer surgery suggests the use of regional anesthesia may reduce cancer recurrence and improve survival. Amide-linked local anesthetics have antiinflammatory properties, although the mechanism of action in this regard is unclear. As inflammatory processes involving Src tyrosine protein kinase and intercellular adhesion molecule-1 are important in tumor growth and metastasis, we hypothesized that amide-linked local anesthetics may inhibit inflammatory Src-signaling involved in migration of adenocarcinoma cells. NCI-H838 lung cancer cells were incubated with tumor necrosis factor-α in absence/presence of ropivacaine, lidocaine, or chloroprocaine (1 nM-100 μM). Cell migration and total cell lysate Src-activation and intercellular adhesion molecule-1 phosphorylation were assessed. The role of voltage-gated sodium-channels in the mechanism of local anesthetic effects was also evaluated. Ropivacaine treatment (100 μM) of H838 cells for 20 min decreased basal Src activity by 62% (P=0.003), and both ropivacaine and lidocaine coadministered with tumor necrosis factor-α statistically significantly decreased Src-activation and intercellular adhesion molecule-1 phosphorylation, whereas chloroprocaine had no such effect. Migration of these cells at 4 h was inhibited by 26% (P=0.005) in presence of 1 μM ropivacaine and 21% by 1 μM lidocaine (P=0.004). These effects of ropivacaine and lidocaine were independent of voltage-gated sodium-channel inhibition. This study indicates that amide-, but not ester-linked, local anesthetics may provide beneficial antimetastatic effects. The observed inhibition of NCI-H838 cell migration by lidocaine and ropivacaine was associated with the inhibition of tumor necrosis factor-α-induced Src-activation and intercellular adhesion molecule-1 phosphorylation, providing the first evidence of a molecular mechanism that appears to be independent of their known role as sodium-channel blockers.
Product Description
Procaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Procaine hydrochloride, ≥97%
Supelco
Procaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Veratridine, ≥90% (HPLC), powder