Merck
  • Vibrational spectral analysis and first hyperpolarizability studies of 1-bromonaphthalene based on ab initio and DFT methods.
Vibrational spectral analysis and first hyperpolarizability studies of 1-bromonaphthalene based on ab initio and DFT methods.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-10-01)
M Arivazhagan, D Anitha Rexalin
ABSTRACT
In this work, the experimental and theoretical vibrational spectra of 1-bromonaphthalene (1-BN) were studied. FTIR and FT Raman spectra were recorded in the region 4000-400 cm(-1) and 3500-50 cm(-1), respectively. The structural and spectroscopic data of the molecule in the ground state were calculated by using ab initio Hartree-Fock and density functional method (B3LYP) with the 6-311++G(d,p) basis set. The vibrational frequencies were calculated and scaled values were compared with experimental FTIR and FT Raman spectra. The observed and calculated frequencies are found to be in good agreement. The complete assignments were performed on the basis of the total energy distribution (TED) of the vibrational modes, calculated with scaled quantum mechanics (SQM) method. The optimized geometric parameters were calculated. The predicted first hyperpolarizability also shows that the molecule might have a reasonably good nonlinear optical (NLO) behaviour. The calculated HOMO-LUMO energy gap reveals that charge transfer occurs within the molecule.
Sigma-Aldrich
1-Bromonaphthalene, 97%
Sigma-Aldrich
1-Bromonaphthalene, ≥95%