Merck
  • Byproduct recovery from reclaimed water reverse osmosis concentrate using lime and soda-ash treatment.
Byproduct recovery from reclaimed water reverse osmosis concentrate using lime and soda-ash treatment.
Water environment research : a research publication of the Water Environment Federation (2010-05-04)
Farah Mohammadesmaeili, Mostafa Kabiri Badr, Morteza Abbaszadegan, Peter Fox
ABSTRACT
Lime and soda-ash softening of reclaimed water reverse osmosis concentrates as a pretreatment step for concentration by seawater reverse osmosis was the focus of this study. The objectives were removal of the potential fouling minerals of calcium, magnesium, and silica by selective precipitation, while producing byproducts with potential resale value. Three different bench-scale lime-soda processes were evaluated. The traditional method produced low-quality magnesium hydroxide [Mg(OH)2] and calcium carbonate (CaCO3) byproducts. A modified process with pre-acidification to eliminate carbonate removed 98 to 99% of calcium and magnesium and produced CaCO3 that was > 94% pure. To prevent the contamination of byproducts with calcium sulfate (CaSO4) in high-sulfate concentrates, a CaSO4 crystallization step was added successfully to the modified process to precipitate CaSO4 before Mg(OH)2 precipitation and produce gypsum that was 92% pure. The modified lime-soda process also removed 94 to 97% silica, 72 to 77% barium, and 95 to 96% strontium, which are known as reverse osmosis membrane foulants.
Sigma-Aldrich
Soda lime, for absorption of carbon dioxide
Sigma-Aldrich
Soda lime, p.a., with indicator, pellets
Sigma-Aldrich
Soda lime, ACS reagent, granular, +100 mesh particle size