Merck
  • Preparation of a highly sensitive enzyme electrode using gold nanoparticles for measurement of pesticides at the ppt level.
Preparation of a highly sensitive enzyme electrode using gold nanoparticles for measurement of pesticides at the ppt level.
Journal of environmental monitoring : JEM (2008-05-02)
Gha-Young Kim, Joonmok Shim, Min-Su Kang, Seung-Hyeon Moon
ABSTRACT
A highly sensitive enzyme electrode was prepared based on gold nanoparticles for measurement of pesticides. Gold nanoparticles of 25-30 nm were synthesized on a glassy carbon electrode by double-pulse technique while the coverage was controlled by applied potential and time. The gold nanoparticles were modified to form a self-assembled monolayer, followed by covalent binding of tyrosinase. The TYR-AuNP-GC electrode was compared with bare GC, AuNP-GC, and modified AuNP-GC and TYR-Au (plate type) electrodes in terms of cyclic voltammetry. The voltammograms well represent the sensitivity of enzymatic oxidation of catechol, substrates for the enzyme activity. The prepared electrode integrated into a continuous flow system and was tested to detect pesticides, such as 2,4-D, atrazine, and ziram. Under the optimized conditions of the flow system, the electrode performed reasonably according to the inhibition mechanism in the concentration range of 0.001-0.5 ng mL(-1). The enhanced performance was attributed to the favored microenvironment for the enzyme activity provided by SAM on gold nanoparticles.
Product Description
Supelco
Ziram, PESTANAL®, analytical standard
Sigma-Aldrich
Zinc dimethyldithiocarbamate, 97%