Merck
  • Relative rotational motion between alpha-Cyclodextrin Derivatives and a stiff axle molecule.
Relative rotational motion between alpha-Cyclodextrin Derivatives and a stiff axle molecule.
The Journal of organic chemistry (2008-03-14)
Dai Nishimura, Tomoya Oshikiri, Yoshinori Takashima, Akihito Hashidzume, Hiroyasu Yamaguchi, Akira Harada
ABSTRACT
Novel [2]rotaxanes bearing alpha-cyclodextrin (alpha-CD) derivatives and a diphenylacetylene axis molecule with trinitrobenzene as a bulky stopper have been prepared to investigate the relative rotary movement of a ring relative to an axis molecule and that of an axis molecule in a ring by NMR techniques. [2]Rotaxanes 2 and 3 were composed of alpha-CD derivatives (2: 6-phenyl-amide-alpha-CD; 3: 6-stilbene-amide-alpha-CD). The protons of alpha-CDs in rotaxanes were thoroughly assigned by the two-dimensional NMR techniques (TOCSY, COSY, ROESY, HMQC, and HMBC). The protons of alpha-CD in rotaxane 1 did not show splitting, whereas the resonance peak shifts and splitting for the corresponding protons of alpha-CD derivatives in rotaxanes 2 and 3 were observed by the shielding and deshielding effects from a diphenylacetylene axis molecule. The splitting of resonance peaks was closely related to the rotary movements of alpha-CDs and an axis molecule. We supposed that alpha-CD in rotaxane 1 rotates freely around a diphenylacetylene axis molecule, and vice versa, whereas the rotary movement of alpha-CD derivatives and the axis molecules of rotaxanes 2 and 3 were restricted by the steric repulsion between the substituent group of alpha-CD and the stopper group of an axis molecule. To estimate the relative rotary movement of CDs and an axis molecule in rotaxanes, the rotational correlation time (tauc) of rotaxanes was measured by 13C NMR. The results indicate that the corresponding rotary movement of the modified alpha-CD and the axis molecules in rotaxanes 2 and 3 depends on the size of the substituent group.
Sigma-Aldrich
Diphenylacetylene, 98%
Pricing and availability is not currently available.