Merck
  • Selective inactivation of eukaryotic beta-galactosidase in assays for inhibitors of HIV-1 TAT using bacterial beta-galactosidase as a reporter enzyme.
Selective inactivation of eukaryotic beta-galactosidase in assays for inhibitors of HIV-1 TAT using bacterial beta-galactosidase as a reporter enzyme.
Analytical biochemistry (1993-11-15)
D C Young, S D Kingsley, K A Ryan, F J Dutko
ABSTRACT
Bacterial beta-galactosidase is one of several reporter enzymes used in studying the transcriptional activity of eukaryotic promoters. Although it is one of the easiest and least expensive enzymes to assay, its use has been limited because of its low sensitivity, which is due in part to endogenous levels of beta-galactosidase in many eukaryotic cells. In this study, we compared the pH and salt requirements, as well as the heat stability, of bacterial and eukaryotic beta-galactosidase in order to identify conditions which would inhibit the beta-galactosidase enzyme endogenous to eukaryotic cells without adversely affecting the activity of either purified bacterial beta-galactosidase or reporter beta-galactosidase produced after transfection of expression vectors into eukaryotic cells. Heat treatment at 50 degrees C for 1 h inactivated the beta-galactosidase activity endogenous to several eukaryotic cell lines by as much as 40-fold without adversely affecting the activity of bacterial beta-galactosidase. This treatment increased the sensitivity of this reporter enzyme and allowed the development of a rapid and quantifiable screening assay for HIV-1 tat inhibitors.
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Pricing and availability is not currently available.