Merck
  • Gastrin treatment stimulates β-cell regeneration and improves glucose tolerance in 95% pancreatectomized rats.
Gastrin treatment stimulates β-cell regeneration and improves glucose tolerance in 95% pancreatectomized rats.
Endocrinology (2011-05-12)
Noèlia Téllez, Géraldine Joanny, Jéssica Escoriza, Marina Vilaseca, Eduard Montanya
ABSTRACT
β-Cell mass reduction is a central aspect in the development of type 1 and type 2 diabetes, and substitution or regeneration of the lost β-cells is a potentially curative treatment of diabetes. To study the effects of gastrin on β-cell mass in rats with 95% pancreatectomy (95%-Px), a model of pancreatic regeneration, rats underwent 95% Px or sham Px and were treated with [15 leu] gastrin-17 (Px+G and S+G) or vehicle (Px+V and S+V) for 15 d. In 95% Px rats, gastrin treatment reduced hyperglycemia (280 ± 52 mg vs. 436 ± 51 mg/dl, P < 0.05), and increased β-cell mass (1.15 ± 0.15 mg)) compared with vehicle-treated rats (0.67 ± 0.15 mg, P < 0.05). Gastrin treatment induced β-cell regeneration by enhancing β-cell neogenesis (increased number of extraislet β-cells in Px+G: 0.42 ± 0.05 cells/mm(2) vs. Px+V: 0.27 ± 0.07 cells/mm(2), P < 0.05, and pancreatic and duodenal homeobox 1 expression in ductal cells of Px+G: 1.21 ± 0.38% vs. Px+V: 0.23 ± 0.10%, P < 0.05) and replication (Px+G: 1.65 ± 0.26% vs. S+V: 0.64 ± 0.14%; P < 0.05). In addition, reduced β-cell apoptosis contributed to the increased β-cell mass in gastrin-treated rats (Px+G: 0.07 ± 0.02%, Px+V: 0.23 ± 0.05%; P < 0.05). Gastrin action on β-cell regeneration and survival increased β-cell mass and improved glucose tolerance in 95% Px rats, supporting a potential role of gastrin in the treatment of diabetes.
Sigma-Aldrich
[Leu15]-Gastrin I human, ≥95% (HPLC)
Sigma-Aldrich
Gastrin II Sulfated, ≥95% (HPLC)