Merck
  • Panobinostat mediated cell death: a novel therapeutic approach for osteosarcoma.
Panobinostat mediated cell death: a novel therapeutic approach for osteosarcoma.
Oncotarget (2018-09-27)
André Wirries, Samir Jabari, Esther P Jansen, Silvia Roth, Elizabeth Figueroa-Juárez, Thaddeus T Wissniowski, Daniel Neureiter, Eckhard Klieser, Philipp Lechler, Steffen Ruchholtz, Detlef K Bartsch, Christoph K Boese, Pietro Di Fazio
ABSTRACT
Osteosarcoma is an aggressive cancer with a poor long term prognosis. Neo-adjuvant poly-chemotherapy followed by surgical resection remains the standard treatment, which is restricted by multi-drug resistance. If first-line therapy fails, disease control and patient survival rate drop dramatically. We aimed to identify alternative apoptotic mechanisms induced by the histone deacetylase inhibitor panobinostat in osteosarcoma cells. Saos-2, MG63 and U2-OS osteosarcoma cell lines, the immortalized human osteoblast line hFOB and the mouse embryo osteoblasts (MC3T3-E1) were treated with panobinostat. Real time viability and FACS confirmed the cytotoxicity of panobinostat. Cell stress/death related factors were analysed by RT-qPCR and western blot. Cell morphology was assessed by electron microscopy. 10 nM panobinostat caused cell viability arrest and death in all osteosarcoma and osteoblast cells. P21 up-regulation was observed in osteosarcoma cells, while over-expression of p73 was restricted to Saos-2 (TP53-/-). Survivin and Bcl-2 were suppressed by panobinostat. Endoplasmic reticulum (ER) stress markers BiP, CHOP, ATF4 and ATF6 were induced in osteosarcoma cells. The un-spliced Xbp was no further detectable after treatment. Autophagy players Beclin1, Map1LC3B and UVRAG transcripts over-expressed after 6 hours. Protein levels of Beclin1, Map1LC3B and p62 were up-regulated at 72 hours. DRAM1 was stable. Electron micrographs revealed the fragmentation and the disappearance of the ER and the statistically significant increase of autophagosome vesiculation after treatment. Panobinostat showed a synergistic suppression of survival and promotion of cell death in osteosarcoma cells. Panobinostat offers new perspectives for the treatment of osteosarcoma and other malignant bone tumours.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-DRAM (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution