Merck
Fibroblast growth factors: at the heart of angiogenesis.
Cell biology international (1995-05-01)
J Slavin
ABSTRACT
Acidic fibroblast growth factor (FGF-1) and basic fibroblast growth factor (FGF-2) are ubiquitous cytokines found in many tissues. They have effects on multiple cell types derived from mesoderm and neuroectoderm, including endothelial cells. In this review the structure and function of the fibroblast growth factor family and its receptors are described. The evidence implicating both FGF-1 and FGF-2 in the control of blood vessel formation is presented and their involvement in normal and pathological angiogenesis during adult life is then described in more detail.
Sigma-Aldrich
FGF-1 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)