
Warfarin Related Compound A

Synonym(s):
3-(2-Hydroxyphenyl)-5-phenyl-2-cyclohexen-1-one, Alice’s ketone
Empirical Formula (Hill Notation):
C18H16O2
CAS Number:
Molecular Weight:
264.32
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2