
Trimethylphenylammonium bromide

Synonym(s):
Phenyltrimethylammonium bromide
Linear Formula:
(CH3)3N(Br)C6H5
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing