
Sumatriptan Succinate Related Compound A

Synonym(s):
3-[2-(Dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl-]methyl]-1H-indol-5-yl]-N-methylmethanesulfonamide succinate salt
Empirical Formula (Hill Notation):
C31H43N5O6S
Molecular Weight:
613.77
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2