
Sertraline hydrochloride Related Compound A

Synonym(s):
1R,4S)-rel-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride
Empirical Formula (Hill Notation):
C17H17Cl2N · HCl
CAS Number:
Molecular Weight:
342.69
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2