
Pentaerythritol tetranitrate solution

Synonym(s):
PETN
Empirical Formula (Hill Notation):
C5H8N4O12
CAS Number:
Molecular Weight:
316.14
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2