
Paeonol

Synonym(s):
2′-Hydroxy-4′-methoxyacetophenone, Paeonol, Resacetophenone 4-O-methyl ether
Linear Formula:
HOC6H3(OCH3)COCH3
CAS Number:
Molecular Weight:
166.17
Products
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing